Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144280.2 | NP_001137752.1 | 11 - 12 | 4221 | 69 | |
NM_166669.4 | NP_726441.1 | 11 - 12 | 4262 | 69 | ||
NM_206213.3 | NP_995935.1 | 11 - 12 | 4655 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |