Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ca-alpha1T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103419.2 | NP_001096889.1 | 22 - 23 | 6853 | 97 | |
NM_001258615.1 | NP_001245544.1 | 21 - 22 | 6778 | 97 | ||
NM_001258616.1 | NP_001245545.1 | 22 - 23 | 6838 | 97 | ||
NM_001272338.1 | NP_001259267.1 | 23 - 24 | 6892 | 97 | ||
NM_132068.3 | NP_572296.2 | 22 - 23 | 6802 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |