Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dlp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079347.3 | NP_524071.2 | 2 - 3 | 952 | 62 | |
NM_206353.2 | NP_996075.2 | 2 - 3 | 952 | 62 | ||
GenBank mRNA | AY052017.1 | - | 2 - 3 | 933 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |