Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: teq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169922.3 | NP_001163393.2 | 5 - 6 | 1662 | 84 | |
NM_140031.3 | NP_648288.2 | 5 - 6 | 1662 | 84 | ||
NM_168315.3 | NP_729452.1 | 5 - 6 | 1662 | 84 | ||
GenBank mRNA | BT132770.1 | - | 5 - 6 | 1660 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |