Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144330.2 | NP_001137802.1 | 4 - 5 | 727 | 90 | |
NM_078771.3 | NP_523495.2 | 3 - 4 | 1062 | 90 | ||
NM_164702.2 | NP_723203.1 | 4 - 5 | 601 | 90 | ||
NM_164703.2 | NP_723204.1 | 4 - 5 | 588 | 90 | ||
NM_164704.2 | NP_723205.1 | 3 - 4 | 578 | 90 | ||
GenBank mRNA | AY069229.1 | - | 3 - 4 | 560 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |