Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078778.5 | NP_523502.1 | 2 - 3 | 804 | 99 | |
GenBank mRNA | BT133499.1 | - | 2 - 3 | 802 | 99 | |
M17230.1 | - | 2 - 3 | 788 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |