Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316387.1 | NP_001303316.1 | 4 - 5 | 2715 | 57 | |
NM_164818.2 | NP_723391.2 | 4 - 5 | 2715 | 57 | ||
GenBank mRNA | AF179360.1 | - | 3 - 4 | 2581 | 57 | |
AJ249178.1 | - | 3 - 4 | 2344 | 57 | ||
AY089558.1 | - | 4 - 5 | 2699 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |