Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201802.2 | NP_001188731.1 | 16 - 17 | 2948 | 82 | |
NM_001201803.2 | NP_001188732.1 | 15 - 16 | 3062 | 82 | ||
NM_001273336.1 | NP_001260265.1 | 15 - 16 | 3062 | 82 | ||
NM_001273337.1 | NP_001260266.1 | 15 - 16 | 3021 | 82 | ||
NM_001273338.1 | NP_001260267.1 | 15 - 16 | 3062 | 82 | ||
NM_135399.5 | NP_609243.3 | 15 - 16 | 2742 | 82 | ||
NM_164830.5 | NP_723407.2 | 15 - 16 | 3062 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |