Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RluA-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273422.1 | NP_001260351.1 | 2 - 3 | 811 | 80 | |
NM_164930.2 | NP_723592.1 | 2 - 3 | 1081 | 80 | ||
NM_164931.2 | NP_723593.1 | 2 - 3 | 1215 | 80 | ||
NM_164932.2 | NP_723594.1 | 2 - 3 | 922 | 80 | ||
GenBank mRNA | BT044578.1 | - | 2 - 3 | 1080 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |