Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: bru2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042891.3 | NP_001036356.1 | 3 - 4 | 1632 | 63 | |
NM_001259071.2 | NP_001246000.1 | 3 - 4 | 1632 | 63 | ||
NM_001273481.1 | NP_001260410.1 | 3 - 4 | 1915 | 63 | ||
NM_001273482.1 | NP_001260411.1 | 3 - 4 | 1539 | 63 | ||
NM_135715.4 | NP_609559.1 | 3 - 4 | 1539 | 63 | ||
NM_165007.3 | NP_723742.2 | 3 - 4 | 1539 | 63 | ||
NM_176025.3 | NP_788039.1 | 3 - 4 | 1632 | 63 | ||
GenBank mRNA | AY069464.1 | - | 3 - 4 | 1506 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |