Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp60D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135812.2 | NP_609656.1 | 2 | 1679 | 65 | |
NM_165043.2 | NP_723818.1 | 1 | 1745 | 65 | ||
GenBank mRNA | AY089262.1 | - | 2 | 1678 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |