Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144354.3 | NP_001137826.1 | 5 - 6 | 1622 | 100 | |
NM_001169504.2 | NP_001162975.1 | 5 - 6 | 1622 | 100 | ||
NM_078843.6 | NP_523567.5 | 4 - 5 | 1595 | 100 | ||
NM_165089.4 | NP_723879.3 | 4 - 5 | 1595 | 100 | ||
GenBank mRNA | BT031256.1 | - | 4 - 5 | 1565 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |