Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273529.2 | NP_001260458.1 | 6 - 7 | 1802 | 107 | |
NM_001316393.1 | NP_001303322.1 | 6 - 7 | 1756 | 107 | ||
NM_165103.3 | NP_723899.1 | 6 - 7 | 1802 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |