Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: CycE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259108.2 | NP_001246037.1 | 4 - 5 | 1986 | 60 | |
NM_057611.5 | NP_476959.1 | 5 - 6 | 2276 | 60 | ||
NM_057612.5 | NP_476960.1 | 3 - 4 | 2276 | 60 | ||
NM_165123.3 | NP_723924.1 | 5 - 6 | 2114 | 60 | ||
NM_165124.3 | NP_723925.1 | 6 - 7 | 2366 | 60 | ||
NM_165125.3 | NP_723926.1 | 5 - 6 | 2123 | 60 | ||
GenBank mRNA | AY069507.2 | - | 5 - 6 | 2116 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |