Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259119.2 | NP_001246048.1 | 6 - 7 | 1011 | 88 | |
NM_001259120.1 | NP_001246049.1 | 6 - 7 | 1166 | 88 | ||
NM_001259121.2 | NP_001246050.1 | 5 - 6 | 961 | 88 | ||
NM_165184.3 | NP_724002.2 | 6 - 7 | 1153 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |