Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038817.1 | NP_001033906.1 | 6 - 7 | 974 | 83 | |
NM_001038818.1 | NP_001033907.1 | 6 - 7 | 1147 | 83 | ||
NM_001273526.1 | NP_001260455.1 | 6 - 7 | 1220 | 83 | ||
NM_001273527.1 | NP_001260456.1 | 2 - 3 | 567 | 83 | ||
NM_078840.5 | NP_523564.1 | 5 - 6 | 1142 | 83 | ||
NM_205988.3 | NP_995710.1 | 6 - 7 | 1238 | 83 | ||
NM_205989.3 | NP_995711.1 | 6 - 7 | 1234 | 83 | ||
GenBank mRNA | AF168467.1 | - | 6 - 7 | 1209 | 83 | |
BT010088.1 | - | 5 - 6 | 1142 | 83 | ||
BT133412.1 | - | 2 - 3 | 513 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |