Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103497.2 | NP_001096967.1 | 10 - 11 | 4786 | 62 | |
NM_078589.4 | NP_511144.2 | 9 - 10 | 4468 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |