Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkcdelta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144719.2 | NP_001138191.1 | 7 - 8 | 1419 | 82 | |
NM_078616.4 | NP_511171.3 | 7 - 8 | 1595 | 82 | ||
GenBank mRNA | BT021275.1 | - | 6 - 7 | 1351 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |