Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CdGAPr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169546.2 | NP_001163017.1 | 6 - 7 | 5598 | 66 | |
NM_001273671.2 | NP_001260600.1 | 7 - 8 | 5673 | 66 | ||
NM_136158.5 | NP_610002.1 | 6 - 7 | 5601 | 66 | ||
GenBank mRNA | AF218776.1 | - | 6 - 7 | 5590 | 66 | |
BT132944.1 | - | 6 - 7 | 5568 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |