Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: koi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273809.1 | NP_001260738.1 | 6 - 7 | 2868 | 62 | |
NM_001273810.1 | NP_001260739.1 | 5 - 6 | 2825 | 62 | ||
NM_136396.4 | NP_610240.4 | 5 - 6 | 2692 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |