Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103953.3 | NP_001097423.1 | 5 - 6 | 1892 | 61 | |
NM_001259540.1 | NP_001246469.1 | 5 - 6 | 2400 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |