Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha68-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273497.1 | NP_001260426.1 | 4 - 5 | 1943 | 67 | |
NM_078836.3 | NP_523560.2 | 4 - 5 | 1876 | 67 | ||
GenBank mRNA | AY089512.1 | - | 4 - 5 | 1851 | 67 | |
U19745.2 | - | 4 - 5 | 1851 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |