Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103768.3 | NP_001097238.1 | 2 - 3 | 1263 | 72 | |
NM_001259293.2 | NP_001246222.1 | 2 - 3 | 1288 | 72 | ||
NM_001273891.1 | NP_001260820.1 | 2 - 3 | 349 | 72 | ||
NM_136634.4 | NP_610478.1 | 2 - 3 | 1289 | 72 | ||
GenBank mRNA | AY060400.1 | - | 2 - 3 | 1258 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |