Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299391.1 | NP_001286320.1 | 3 - 4 | 888 | 117 | |
NM_136859.3 | NP_610703.3 | 3 - 4 | 1169 | 117 | ||
GenBank mRNA | AY058444.1 | - | 3 - 4 | 874 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |