Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cals | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272157.1 | NP_001259086.1 | 10 - 11 | 2933 | 109 | |
NM_001297793.1 | NP_001284722.1 | 10 - 11 | 2936 | 109 | ||
NM_079893.3 | NP_524632.1 | 10 - 11 | 2933 | 109 | ||
NM_166823.2 | NP_726646.1 | 10 - 11 | 2936 | 109 | ||
GenBank mRNA | AJ289018.1 | - | 10 - 11 | 2899 | 109 | |
AY121644.1 | - | 10 - 11 | 2901 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |