Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: norpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014720.2 | NP_001014720.1 | 9 - 10 | 2156 | 58 | |
NM_001014721.3 | NP_001014721.1 | 8 - 9 | 1694 | 58 | ||
NM_001169190.2 | NP_001162661.1 | 10 - 11 | 2425 | 58 | ||
NM_001297931.1 | NP_001284860.1 | 9 - 10 | 1819 | 58 | ||
NM_080330.4 | NP_525069.2 | 8 - 9 | 1694 | 58 | ||
NM_167008.2 | NP_726925.1 | 9 - 10 | 2156 | 58 | ||
GenBank mRNA | AF181641.1 | - | 8 - 9 | 1677 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |