Search Thermo Fisher Scientific
Gene Symbol: yellow-d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137944.4 | NP_611788.1 | 1 - 2 | 264 | 67 | |
GenBank mRNA | AY121706.1 | - | 1 - 2 | 261 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |