Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: JYalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110622.3 | NP_001104092.2 | 5 - 6 | 2954 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |