Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169877.2 | NP_001163348.1 | 2 - 3 | 877 | 99 | |
NM_001169878.2 | NP_001163349.1 | 2 - 3 | 877 | 99 | ||
NM_001274488.1 | NP_001261417.1 | 2 - 3 | 989 | 99 | ||
NM_001274489.1 | NP_001261418.1 | 2 - 3 | 585 | 99 | ||
NM_168090.3 | NP_729009.2 | 2 - 3 | 877 | 99 | ||
NM_168091.3 | NP_729010.1 | 2 - 3 | 989 | 99 | ||
NM_168092.3 | NP_729011.1 | 2 - 3 | 585 | 99 | ||
GenBank mRNA | BT120208.1 | - | 2 - 3 | 985 | 99 | |
BT120209.1 | - | 2 - 3 | 985 | 99 | ||
FJ172423.1 | - | 2 - 3 | 873 | 99 | ||
FJ172424.1 | - | 2 - 3 | 873 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |