Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274977.1 | NP_001261906.1 | 5 - 6 | 4191 | 98 | |
NM_001274978.1 | NP_001261907.1 | 6 - 7 | 4203 | 98 | ||
NM_080497.5 | NP_536745.4 | 5 - 6 | 4179 | 98 | ||
NM_080498.3 | NP_536746.1 | 5 - 6 | 4191 | 98 | ||
NM_168640.2 | NP_730088.1 | 5 - 6 | 4174 | 98 | ||
NM_168641.1 | NP_730089.1 | 4 - 5 | 4405 | 98 | ||
GenBank mRNA | AY095048.1 | - | 5 - 6 | 4189 | 98 | |
BT009972.1 | - | 5 - 6 | 4179 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |