Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259855.2 | NP_001246784.1 | 10 - 11 | 2324 | 80 | |
NM_001259856.2 | NP_001246785.1 | 10 - 11 | 2580 | 80 | ||
NM_001259857.2 | NP_001246786.1 | 10 - 11 | 2327 | 80 | ||
NM_168646.3 | NP_730099.2 | 9 - 10 | 1909 | 80 | ||
GenBank mRNA | BT133053.1 | - | 9 - 10 | 1897 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |