Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GluRIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274692.1 | NP_001261621.2 | 12 - 13 | 2656 | 72 | |
NM_001274693.1 | NP_001261622.1 | 12 - 13 | 2656 | 72 | ||
NM_001316480.1 | NP_001303409.1 | 12 - 13 | 2656 | 72 | ||
NM_080016.4 | NP_524755.2 | 12 - 13 | 2656 | 72 | ||
GenBank mRNA | BT150463.1 | - | 12 - 13 | 2655 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |