Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103495.5 | NP_001096965.1 | 6 - 7 | 2070 | 94 | |
NM_001258717.1 | NP_001245646.1 | 7 - 8 | 3590 | 94 | ||
NM_001258718.3 | NP_001245647.1 | 7 - 8 | 3375 | 94 | ||
NM_001258719.2 | NP_001245648.1 | 6 - 7 | 3156 | 94 | ||
NM_001258720.3 | NP_001245649.2 | 5 - 6 | 2402 | 94 | ||
NM_001272568.2 | NP_001259497.1 | 5 - 6 | 1862 | 94 | ||
NM_001272569.1 | NP_001259498.1 | 6 - 7 | 2328 | 94 | ||
NM_001298244.1 | NP_001285173.1 | 5 - 6 | 1853 | 94 | ||
NM_132614.6 | NP_572842.1 | 5 - 6 | 2411 | 94 | ||
GenBank mRNA | AF239667.1 | - | 4 - 5 | 805 | 94 | |
BT120384.2 | - | 4 - 5 | 747 | 94 | ||
BT125663.1 | - | 4 - 5 | 738 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |