Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RyR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259277.1 | NP_001246206.1 | 17 - 18 | 8826 | 85 | |
NM_001259278.2 | NP_001246207.1 | 18 - 19 | 9024 | 85 | ||
NM_001259279.2 | NP_001246208.1 | 17 - 18 | 8809 | 85 | ||
NM_001259280.1 | NP_001246209.1 | 17 - 18 | 9089 | 85 | ||
NM_001259281.2 | NP_001246210.1 | 18 - 19 | 9066 | 85 | ||
NM_001259282.2 | NP_001246211.1 | 16 - 17 | 8871 | 85 | ||
NM_057643.4 | NP_476991.1 | 17 - 18 | 8833 | 85 | ||
NM_057644.4 | NP_476992.1 | 17 - 18 | 8791 | 85 | ||
NM_057645.3 | NP_476993.1 | 17 - 18 | 8868 | 85 | ||
NM_057646.4 | NP_476994.1 | 17 - 18 | 8791 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |