Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058115.5 | NP_477463.1 | 1 - 2 | 337 | 70 | |
GenBank mRNA | AJ243836.2 | - | 1 - 2 | 306 | 70 | |
AJ276419.1 | - | 1 - 2 | 311 | 70 | ||
AY118419.1 | - | 1 - 2 | 312 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |