Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103767.2 | NP_001097237.1 | 6 - 7 | 4031 | 76 | |
NM_001169624.1 | NP_001163095.1 | 5 - 6 | 3380 | 76 | ||
NM_001201966.2 | NP_001188895.1 | 8 - 9 | 3975 | 76 | ||
NM_001201967.1 | NP_001188896.1 | 6 - 7 | 3590 | 76 | ||
NM_001201968.3 | NP_001188897.3 | 7 - 8 | 3495 | 76 | ||
NM_001259291.2 | NP_001246220.1 | 7 - 8 | 3436 | 76 | ||
NM_001259292.2 | NP_001246221.1 | 7 - 8 | 3567 | 76 | ||
NM_001299312.1 | NP_001286241.1 | 8 - 9 | 4087 | 76 | ||
NM_001299313.1 | NP_001286242.1 | 7 - 8 | 3157 | 76 | ||
NM_001299314.1 | NP_001286243.1 | 7 - 8 | 3496 | 76 | ||
NM_001299315.1 | NP_001286244.1 | 7 - 8 | 3490 | 76 | ||
NM_176110.2 | NP_788290.1 | 6 - 7 | 3226 | 76 | ||
NM_176111.2 | NP_788291.1 | 7 - 8 | 3877 | 76 | ||
NM_176112.2 | NP_788292.1 | 6 - 7 | 3348 | 76 | ||
NM_176113.3 | NP_788293.2 | 7 - 8 | 3999 | 76 | ||
NM_176114.3 | NP_788294.1 | 3 - 4 | 1096 | 76 | ||
GenBank mRNA | BT133091.1 | - | 7 - 8 | 3840 | 76 | |
BT133361.1 | - | 3 - 4 | 1105 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |