Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: abd-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170161.2 | NP_001163632.1 | 3 - 4 | 1856 | 72 | |
NM_001260216.2 | NP_001247145.1 | 3 - 4 | 2153 | 72 | ||
NM_057345.5 | NP_476693.1 | 4 - 5 | 1755 | 72 | ||
NM_169733.5 | NP_732176.1 | 2 - 3 | 1232 | 72 | ||
GenBank mRNA | BT016031.1 | - | 3 - 4 | 2143 | 72 | |
X54453.1 | - | 1018 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |