Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001363924.1 | NP_001350861.1 | 2 - 3 | 512 | 91 | |
NM_058155.5 | NP_477503.1 | 2 - 3 | 512 | 91 | ||
GenBank mRNA | BT150358.1 | - | 2 - 3 | 512 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |