Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@f0ecd33
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Gene Expression
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm02136018_g1
See other CBP20 GE Assays ›
Gene Symbol:
Cbp20
Gene Name:
cap binding protein 20
Gene Aliases:
CBP20, CG12357, Dmel\CG12357, Dmel_CG12357, cbp20
Chromosome Location:
Chr.3R: 18164661 - 18165439 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0022943
Assay ID
Dm02136018_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
Displaying...
Build 9.4 Dros Chr.3R: Dm02136018_g1
×
Created with Raphaël 2.1.2
CG7218
Cbp20
Prx5
CG7215
NM_00130
0456.1
NM_14242
1.4
NM_07967
2.3
NM_00103
2020.3
NM_14242
2.5
AY058285
.1
AY070578
.1
AJ238969
.1
BT003526
.1
AY051983
.1
13,987,2
00
13,987,3
00
13,987,4
00
13,987,5
00
13,987,6
00
13,987,7
00
13,987,8
00
13,987,9
00
13,988,0
00
13,988,1
00
13,988,2
00
13,988,3
00
13,988,4
00
13,988,5
00
13,988,6
00
13,988,7
00
13,988,8
00
13,988,9
00
13,989,0
00
13,989,1
00
13,989,2
00
13,989,3
00
13,989,4
00
13,989,5
00
13,989,6
00
13,989,7
00
13,989,8
00
13,989,9
00
13,990,0
00
13,990,1
00
13,990,2
00
13,990,3
00
13,990,4
00
13,990,5
00
13,990,6
00
13,990,7
00
13,990,8
00
13,990,9
00
13,991,0
00
13,991,1
00
13,991,2
00
13,991,3
00
13,991,4
00
13,991,5
00
13,991,6
00
13,991,7
00
13,991,8
00
13,991,9
00
13,992,0
00
13,992,1
00
13,992,2
00
13,992,3
00
13,992,4
00
13,992,5
00
13,992,6
00
13,992,7
00
13,992,8
00
13,992,9
00
13,993,0
00
13,993,1
00
13,993,2
00
13,993,3
00
13,993,4
00
13,993,5
00
13,993,6
00
13,993,7
00
13,993,8
00
13,993,9
00
13,994,0
00
13,994,1
00
13,994,2
00
13,994,3
00
13,994,4
00
Show all RefSeq and GenBank
Back To Top
Assay Details
Gene Symbol: Cbp20
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_079672.3
NP_524396.1
1 - 2
121
101
GenBank mRNA
AJ238969.1
-
1 - 2
86
101
AY070578.1
-
1 - 2
119
101
KX531599.1
-
1 - 2
53
101
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Drosophila melanogaster
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
RNA interference
production of siRNA involved in RNA interference
primary miRNA processing
gene silencing by miRNA
negative regulation of viral genome replication
mRNA cis splicing, via spliceosome
RNA cap binding
mRNA binding
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c659bcd8-gmtsf:80/100.66.75.26:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline