Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: r-l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079703.3 | NP_524427.1 | 1 - 2 | 251 | 77 | |
GenBank mRNA | AY058714.1 | - | 1 - 2 | 229 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |