Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cont | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141204.4 | NP_649461.2 | 5 - 6 | 3888 | 62 | |
GenBank mRNA | AY229991.1 | - | 4 - 5 | 3611 | 62 | |
BT021333.1 | - | 5 - 6 | 3884 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |