Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sra-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_142190.3 | NP_650447.1 | 4 - 5 | 2569 | 90 | |
GenBank mRNA | AY017343.1 | - | 4 - 5 | 2556 | 90 | |
AY029211.1 | - | 4 - 5 | 2556 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |