Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382180.1 | NP_001368946.1 | 4 - 5 | 1549 | 74 | |
NM_143312.4 | NP_651569.1 | 4 - 5 | 1709 | 74 | ||
NM_170336.3 | NP_733215.1 | 4 - 5 | 1870 | 74 | ||
NM_170337.3 | NP_733216.1 | 4 - 5 | 1549 | 74 | ||
NM_170338.3 | NP_733217.1 | 5 - 6 | 1718 | 74 | ||
NM_170339.3 | NP_733218.1 | 5 - 6 | 1878 | 74 | ||
NM_170340.2 | NP_733219.1 | 4 - 5 | 1962 | 74 | ||
GenBank mRNA | AF145696.1 | - | 4 - 5 | 1836 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |