Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300660.1 | NP_001287589.1 | 6 - 7 | 2268 | 62 | |
NM_001300661.1 | NP_001287590.1 | 6 - 7 | 2222 | 62 | ||
NM_143459.3 | NP_651716.1 | 6 - 7 | 2222 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |