Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lerp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276067.1 | NP_001262996.1 | 9 - 10 | 1253 | 112 | |
NM_170288.3 | NP_733167.2 | 9 - 10 | 1268 | 112 | ||
NM_170289.3 | NP_733168.2 | 9 - 10 | 1289 | 112 | ||
NM_170290.3 | NP_733169.2 | 9 - 10 | 1289 | 112 | ||
GenBank mRNA | AY684632.2 | - | 9 - 10 | 1268 | 112 | |
BT021416.1 | - | 9 - 10 | 1233 | 112 |
Set Membership: |
![]() ![]() ![]() |