Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079583.3 | NP_524307.1 | 2 - 3 | 469 | 124 | |
GenBank mRNA | KX531658.1 | - | 1 - 2 | 321 | 124 |
Set Membership: |
![]() ![]() ![]() ![]() |