Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079912.4 | NP_524651.2 | 2 | 4758 | 67 | |
NM_168882.2 | NP_730596.1 | 2 | 4203 | 67 | ||
NM_168883.1 | NP_730597.1 | 2 | 4092 | 67 | ||
GenBank mRNA | AF195498.1 | - | 2 | 4744 | 67 | |
AF250842.1 | - | 2 | 4743 | 67 | ||
AY069579.1 | - | 2 | 4739 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |