Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042795.3 | NP_001036260.3 | 3 - 4 | 643 | 104 | |
NM_001144679.2 | NP_001138151.2 | 4 - 5 | 555 | 104 | ||
GenBank mRNA | BT083378.1 | - | 4 - 5 | 555 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |