Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mRpS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043186.2 | NP_001036651.1 | 1 - 2 | 252 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |