Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001845.2 | NP_001001845.2 | 57 - 58 | 12200 | 100 | |
XM_005165511.4 | XP_005165568.1 | 57 - 58 | 12319 | 100 | ||
XM_009301753.3 | XP_009300028.1 | 58 - 59 | 12341 | 100 | ||
GenBank mRNA | AY540131.1 | - | 57 - 58 | 12204 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |